Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5114927

Description
Antibody detects endogenous levels of total CACNA1B.
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications
CACNA1B | |
Polyclonal | |
Unconjugated | |
CACNA1B | |
alpha(1B); AW050276; AW060892; AW822256; BIII; Brain calcium channel III; Cach5; Cacna1b; Cacnl1a5; CACNN; calcium channel alpha12.2 subunit; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 5; calcium channel, voltage-dependent, L type, alpha 1B subunit; calcium channel, voltage-dependent, N type, alpha 1B subunit; calcium voltage-gated channel subunit alpha1 B; Cav2.2; Cav2.2 voltage-gated Ca2+ channel; Cchn1a; DYT23; voltage gated N-type calcium channel Ca(v)2.2; voltage-dependent N-type calcium channel subunit alpha-1B; voltage-gated calcium channel alpha subunit Cav2.2; voltage-gated calcium channel subunit alpha Cav2.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12287, 257648, 774 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O55017, Q00975, Q02294 | |
CACNA1B | |
A synthesized peptide derived from human CACNA1B(Accession Q00975), corresponding to amino acid residues G264-E314. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction