Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA1E Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | CACNA1E |
---|---|
Dilution | Western Blot 5 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CACNA1E Polyclonal antibody specifically detects CACNA1E in Human samples. It is validated for Western BlotSpecifications
CACNA1E | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3 | |
Synthetic peptide derived from the rat alpha1E calcium channel conjugated to KLH (Uniprot: Q07652) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 5 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
777 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title