Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA1F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584778 Supplier Invitrogen™ Supplier No. PA584778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCEDLPIPGT YHRGRNSGPN RAQGSWATPP QRGRLLYAPL LLVEEGAAGE GYLGRSSGPL RTFTCLHVPG THSDPSH Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 82%.

Voltage-sensitive calcium channels (VSCC) mediate the entry of calcium ions into excitable cells and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. The isoform alpha-1F gives rise to L-type calcium currents. Long-lasting (L-type) calcium channels belong to the 'high-voltage activated' (HVA) group. They are blocked by dihydropyridines (DHP), phenylalkylamines, benzothiazepines, and by omega-agatoxin-IIIA (omega-Aga-IIIA). They are however insensitive to omega-conotoxin-GVIA (omega-CTx-GVIA) and omega-agatoxin-IVA (omega-Aga-IVA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA1F
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CACNA1F
Gene Accession No. O60840
Gene Alias A930034B14; AIED; Cacna1f; CACNAF1; calcium channel, voltage-dependent, alpha 1F subunit; calcium channel, voltage-dependent, L type, alpha 1F subunit; calcium voltage-gated channel subunit alpha1 F; Cav1.4; Cav1.4alpha1; COD3; COD4; CORDX; CORDX3; CSNB2; CSNB2A; CSNBX2; JM8; JMC8; L-type dihydropyridine-sensitive calcium channel alpha-1f subunit; nerg1; nob2; OA2; Sfc17; Voltage-dependent L-type calcium channel subunit alpha-1F; voltage-gated calcium channel subunit alpha Cav1.4
Gene Symbols CACNA1F
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CACNA1F. Recombinant protein control fragment (Product #RP-108540).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.