Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA1G Antibody (S178A-9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259322
Description
CACNA1G Monoclonal specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CACNA1G | |
Monoclonal | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
CACNA1G | |
Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1 | |
100 μg | |
Primary | |
Detects <200kDa. Does not cross-react with Cav3.2. | |
Store at -20C. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S178A-9 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit | |
Mouse | |
Protein G purified | |
RUO | |
8913 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction