Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA1G Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2781R

Description
Voltage-dependent Ca2+ channels mediate Ca2+ entry into excitable cells in response to membrane depolarization, and they are involved in a variety of Ca2+-dependent processes, including muscle contraction, hormone or neurotransmitter release and gene expression. Calcium channels are highly diverse, multimeric complexes composed of an alpha-1 subunit, an intracellular beta subunit, a disulfide linked alpha-2/delta subunit and a transmembrane gamma subunit. Ca2+ currents are characterized on the basis of their biophysical and pharmacologic properties and include L-, N-, T-, P-, Q-, and R- types. T-type Ca2+ currents are activated and inactivated more rapidly and at more negative membrane potentials than other Ca2+ current types. T-type Ca2+ channels enhance odor sensitivity by lowering the threshold of spike generation in olfactory receptor cells (ORCs).Specifications
CACNA1G | |
Polyclonal | |
Unconjugated | |
CACNA1G | |
[a]1G; a1G; alpha-1G; Ca(V)T.1; CACNA1G; calcium channel, voltage-dependent, T type, alpha 1G subunit; calcium voltage-gated channel subunit alpha1 G; CaV T1; Cav3 1; cav3 1c; Cav3.1; Cav3.1c; Cav3.1d; KIAA1123; MGC117234; mKIAA1123; NBR13; SCA42; voltage-dependent calcium channel alpha 1G subunit, isoform 11; voltage-dependent T-type calcium channel alpha 1G subunit; Voltage-dependent T-type calcium channel subunit alpha-1G; Voltage-gated calcium channel subunit alpha Cav3.1 | |
Rabbit | |
Protein A | |
RUO | |
8913 | |
-20°C | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O43497 | |
CACNA1G | |
KLH conjugated synthetic peptide derived from human CACNA1G/Cav31. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction