Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA1I Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144944 Supplier Invitrogen™ Supplier No. PA5144944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHDKQEVQLA ETEAFSLNSD RSSSILLGDD LSLEDPTACP PGRKDSKGEL DPPEPMRVGD LGECFFPLSS TAVSPDPENF LCEMEEIPFN PVR Antigen sequence identity: Mouse 78%; Rat 26%.

CaV3.3 encodes the pore-forming alpha subunit of a voltage gated calcium channel. The encoded protein is a member of a subfamily of calcium channels referred to as is a low voltage-activated, T-type, calcium channel. The channel encoded by CaV3.3 is characterized by a slower activation and inactivation compared to other T-type calcium channels. CaV3.3 may be involved in calcium signaling in neurons. Alternate splicing results in multiple transcript variants of CaV3.3. Voltage-gated calcium channels (CaV) are present in the membrane of most excitable cells and mediate calcium influx in response to depolarization, an proteins such as CaV3.3 regulate intracellular processes such as contraction, secretion, neurotransmission and gene expression. Diseases associated with CACNA1I include Childhood Absence Epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA1I
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CACNA1I
Gene Accession No. Q9P0X4
Gene Alias Ca(v)3.3; CACNA1I; calcium channel, voltage-dependent, alpha 1I subunit; calcium channel, voltage-dependent, T type, alpha 1I subunit; calcium voltage-gated channel subunit alpha1 I; Cav3.3; CaVT.3; hypothetical gene supported by NM_020084; KIAA1120; low voltage-activated T-type calcium channel alpha-1 subunit (CACNA1I); low-voltage-activated calcium channel alpha13.3; RP1-172B20.4; voltage-dependent calcium channel; voltage-dependent T-type calcium channel subunit alpha-1I; Voltage-gated calcium channel subunit alpha Cav3.3
Gene Symbols CACNA1I
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1842-1934. Recombinant protein control fragment (Product #RP-110066).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.