Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CACNA2D1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518731
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
CACNA2D1 | |
Polyclonal | |
Unconjugated | |
Cacna2d1 | |
Ca(v)alpha2delta1; Cacna2; Cacna2d1; CACNL2A; Calcium channel subunit alpha 2 delta (dihydropyridine - sensitive L-type); calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; calcium channel, voltage-dependent, alpha2/delta subunit 1; calcium voltage-gated channel auxiliary subunit alpha2delta 1; CCHL2A; CCHLA2; DHP; DHSCCA; dihydropryridine-sensitive calcium channel alpha-2 subunit; dihydropyridine-sensitive calcium channel alpha 2; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; H_DJ0560O14.1; LINC01112; lncRNA-N3; MHS3; unknown protein; voltage-dependent calcium channel alpha-2 delta subunit precursor; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1; WUGSC:H_DJ0560O14.1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
781 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P54289 | |
Cacna2d1 | |
Synthetic peptide sequence (QEIFNKYNKDKKVR) corresponding to the internal amino acids of CACNA2D1 (aa 370-384). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction