Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582359 Supplier Invitrogen™ Supplier No. PA582359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFLLSLTFPR LLEAVEMEDD DFTASLSKQS CITEQTQYFF DNDSKSFSGV LDCGNCSRIF HGEKLMNTNL IFIMVESKGT CPCDTRLLIQ AEQTSDGPNP CDMVKQPRYR KGP Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.

This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cacna2d1
Gene Accession No. P54289
Gene Alias Ca(v)alpha2delta1; Cacna2; Cacna2d1; CACNL2A; Calcium channel subunit alpha 2 delta (dihydropyridine - sensitive L-type); calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; calcium channel, voltage-dependent, alpha2/delta subunit 1; calcium voltage-gated channel auxiliary subunit alpha2delta 1; CCHL2A; CCHLA2; DHP; DHSCCA; dihydropryridine-sensitive calcium channel alpha-2 subunit; dihydropyridine-sensitive calcium channel alpha 2; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; H_DJ0560O14.1; LINC01112; lncRNA-N3; MHS3; unknown protein; voltage-dependent calcium channel alpha-2 delta subunit precursor; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1; WUGSC:H_DJ0560O14.1
Gene Symbols Cacna2d1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CACNA2D1. Recombinant protein control fragment (Product #RP-89931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.