Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587762 Supplier Invitrogen™ Supplier No. PA587762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPWPGCGPHP GPGTRRPTSG PPRPLWLLLP LLPLLAAPGA SAYSFPQQHT MQHWARRLEQ EVDGVMRIFG GVQQLREIYK DNRNLFEVQE NEPQKLVEKV AGDIESLLDR KVQALKRLAD AAENFQKAHR WQDNIKEEDI VYYDAKADAE LDDPESEDVE RGSKASTLRL DFIEDPNFKN K; Positive Samples: Mouse brain, Rat heart, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.

This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CACNA2D2
Gene Accession No. Q6PHS9, Q8CFG6
Gene Alias a2d2; alpha 2 delta calcium channel subunit; CACNA2D; Cacna2d2; calcium channel, voltage dependent, alpha2/delta subunit 2; calcium channel, voltage-dependent, alpha 2/delta subunit 2; calcium voltage-gated channel auxiliary subunit alpha2delta 2; Cav alpha2/delta2; du; ducky; gene 26; KIAA0558; mKIAA0558; Protein ducky; td; torpid; Voltage-dependent calcium channel subunit alpha-2/delta-2; Voltage-dependent calcium channel subunit alpha-2-2; Voltage-dependent calcium channel subunit delta-2; Voltage-gated calcium channel subunit alpha-2/delta-2
Gene Symbols CACNA2D2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human CACNA2D2 (NP_0011675221).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300992, 56808
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.