Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526743
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526743 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526743 Supplier Invitrogen™ Supplier No. PA526743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

CACNA2D3 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CACNA2D3
Gene Accession No. Q8IZS8
Gene Alias alpha 2 delta-3; alpha2delta3; Cacna2d3; Cacnad3; calcium channel alpha2-delta3 subunit; calcium channel, voltage dependent, alpha2/delta subunit 3; calcium channel, voltage-dependent, alpha 2/delta 3 subunit; calcium channel, voltage-dependent, alpha 2/delta subunit 3; calcium channel, voltage-dependent, alpha2/delta subunit 3; calcium voltage-gated channel auxiliary subunit alpha2delta 3; CaV alpha2delta3; HSA272268; Voltage-dependent calcium channel subunit alpha-2/delta-3; Voltage-dependent calcium channel subunit alpha-2-3; Voltage-dependent calcium channel subunit delta-3; Voltage-gated calcium channel subunit alpha-2/delta-3
Gene Symbols CACNA2D3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1026-1053 amino acids from the C-terminal region of human CACNA2D3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.