Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587802 Supplier Invitrogen™ Supplier No. PA587802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYDYQAMCRA NKESSDGAHG LLDPYNAFLS AVKWIMTELV LFLVEFNLCS WWHSDMTAKA QKLKQTLEPC DTEYPAFVSE RTIKETTGNI ACEDCSKSFV IQQIPSSNLF MVVVDSSCLC ESVAPITMAP IEIRYNESLK C; Positive Samples: U-87 MG, HeLa, Hep G2, Mouse brain, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.

CACNA2D3 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CACNA2D3
Gene Accession No. Q8CFG5, Q8IZS8, Q9Z1L5
Gene Alias alpha 2 delta-3; alpha2delta3; Cacna2d3; Cacnad3; calcium channel alpha2-delta3 subunit; calcium channel, voltage dependent, alpha2/delta subunit 3; calcium channel, voltage-dependent, alpha 2/delta 3 subunit; calcium channel, voltage-dependent, alpha 2/delta subunit 3; calcium channel, voltage-dependent, alpha2/delta subunit 3; calcium voltage-gated channel auxiliary subunit alpha2delta 3; CaV alpha2delta3; HSA272268; Voltage-dependent calcium channel subunit alpha-2/delta-3; Voltage-dependent calcium channel subunit alpha-2-3; Voltage-dependent calcium channel subunit delta-3; Voltage-gated calcium channel subunit alpha-2/delta-3
Gene Symbols CACNA2D3
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12294, 306243, 55799
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.