Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556889 Supplier Invitrogen™ Supplier No. PA556889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NDFINIIAYN DYVHYIEPCF KGILVQADRD NREHFKLLVE ELMVKGVGVV DQALREAFQI LKQFQEAKQG SLCNQAIM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

CACNA2D4 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cacna2d4
Gene Accession No. Q7Z3S7
Gene Alias 5730412N02Rik; BE686333; Cacna2d4; calcium channel, voltage-dependent, alpha 2/delta subunit 4; calcium voltage-gated channel auxiliary subunit alpha2delta 4; LOW QUALITY PROTEIN: voltage-dependent calcium channel subunit alpha-2/delta-4; RCD4; RGD1562038; Voltage-dependent calcium channel subunit alpha-2/delta-4; Voltage-dependent calcium channel subunit alpha-2-4; Voltage-dependent calcium channel subunit delta-4; voltage-gated calcium channel alpha(2)delta-4 subunit; voltage-gated calcium channel subunit alpha-2/delta-4
Gene Symbols Cacna2d4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CACNA2D4. Recombinant protein control fragment (Product #RP-95799).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.