Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA2D4 Polyclonal Antibody
CACNA2D4 Polyclonal Antibody
Supplier: Thermo Scientific PA570453
Description
CACNA2D4 Polyclonal Antibody for Western Blot

Specifications
CACNA2D4 | |
Polyclonal | |
Unconjugated | |
Cacna2d4 | |
5730412N02Rik; BE686333; Cacna2d4; calcium channel, voltage-dependent, alpha 2/delta subunit 4; calcium voltage-gated channel auxiliary subunit alpha2delta 4; LOW QUALITY PROTEIN: voltage-dependent calcium channel subunit alpha-2/delta-4; RCD4; RGD1562038; Voltage-dependent calcium channel subunit alpha-2/delta-4; Voltage-dependent calcium channel subunit alpha-2-4; Voltage-dependent calcium channel subunit delta-4; voltage-gated calcium channel alpha(2)delta-4 subunit; voltage-gated calcium channel subunit alpha-2/delta-4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
319734 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5RJF7 | |
Cacna2d4 | |
synthetic peptide directed towards the middle region of human CACNA2D4 (aa 313-362). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction