Learn More
Invitrogen™ CACNB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596747
Description
Immunogen sequence: ATAALAASPA PVSNLQGPYL ASGDQPLERA TGEHASMHEY PGELGQPPGL YPSSHPPGRA GTLRALSRQD TFDADTPGSR NSAYTELGDS CVDMETDPSE GPGLGDPAGG GTPPARQGSW EDEEEDYEEE LTDNRNRGRN KARYCAEGGG PVLGRNKNEL EGWGRGVYIR; Positive Samples: HepG2; Cellular Location: Cell membrane, Cytoplasmic side, Peripheral membrane protein, sarcolemma.
Calcium Channel 1 is a protein encoded by the CACNB1 gene and belongs to the calcium channel beta subunit family. Calcium Channel 1 plays an important role in the calcium channel by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Calcium Channel 1 is involved in modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Diseases associated with Calcium Channel 1 protein dysfunction include malignant hyperthermia susceptibility.
Specifications
CACNB1 | |
Polyclonal | |
Unconjugated | |
Cacnb1 | |
accessory voltage-dependent L-type calcium channel subunit; skeletal muscle L-type calcium channel beta1a subunit; brain calcium channel beta 1 subunit; CAB1; Cacnb1; Cacnlb1; calcium channel beta 1 subunit; calcium channel beta1a subunit; calcium channel beta-subunit; calcium channel voltage-dependent subunit beta 1; calcium channel, L type, beta 1 polypeptide; calcium channel, voltage-dependent, beta 1 subunit; calcium channel, voltage-dependent, beta 1 subunit isoform 2; calcium voltage-gated channel auxiliary subunit beta 1; Cchb1; CCHLB1; dihydropyridine receptor b1; dihydropyridine receptor beta 1a subunit; dihydropyridine receptor beta 1c subunit; dihydropyridine-sensitive calcium channel beta-subunit; dihydropyridine-sensitive L-type, calcium channel beta-1 subunit; MGC41896; red; relaxed; voltage-dependent calcium channel beta1 subunit; voltage-dependent calcium channel beta1B subunit; voltage-dependent calcium channel beta1C subunit; voltage-dependent calcium channel L-type beta 1 isoform a; voltage-dependent calcium channel L-type beta 1 isoform c; voltage-dependent L-type calcium channel subunit beta-1; zgc:91982 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12295, 50688, 782 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.66 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54283, Q02641, Q8R3Z5 | |
Cacnb1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 429-598 of human CACNB1 (NP_0007143). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.