Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528900 Supplier Invitrogen™ Supplier No. PA528900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain, rat brain. Predicted reactivity: Mouse (97%), Rat (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Calcium Channel 1 is a protein encoded by the CACNB1 gene and belongs to the calcium channel beta subunit family. Calcium Channel 1 plays an important role in the calcium channel by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Calcium Channel 1 is involved in modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Diseases associated with Calcium Channel 1 protein dysfunction include malignant hyperthermia susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Cacnb1
Gene Accession No. P54283, Q02641, Q8R3Z5
Gene Alias accessory voltage-dependent L-type calcium channel subunit; skeletal muscle L-type calcium channel beta1a subunit; brain calcium channel beta 1 subunit; CAB1; Cacnb1; Cacnlb1; calcium channel beta 1 subunit; calcium channel beta1a subunit; calcium channel beta-subunit; calcium channel voltage-dependent subunit beta 1; calcium channel, L type, beta 1 polypeptide; calcium channel, voltage-dependent, beta 1 subunit; calcium channel, voltage-dependent, beta 1 subunit isoform 2; calcium voltage-gated channel auxiliary subunit beta 1; Cchb1; CCHLB1; dihydropyridine receptor b1; dihydropyridine receptor beta 1a subunit; dihydropyridine receptor beta 1c subunit; dihydropyridine-sensitive calcium channel beta-subunit; dihydropyridine-sensitive L-type, calcium channel beta-1 subunit; MGC41896; red; relaxed; voltage-dependent calcium channel beta1 subunit; voltage-dependent calcium channel beta1B subunit; voltage-dependent calcium channel beta1C subunit; voltage-dependent calcium channel L-type beta 1 isoform a; voltage-dependent calcium channel L-type beta 1 isoform c; voltage-dependent L-type calcium channel subunit beta-1; zgc:91982
Gene Symbols Cacnb1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cav beta 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12295, 50688, 782
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.