Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728438 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8438 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8438 Supplier Proteintech Group Inc Supplier No. 130391AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The expression of multiple classes of voltage dependent calcium channels (VDCC′s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC′s exist before birth which is consistent with a role in migration, most N-Type VDCC′s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.

Specifications

Antigen CACNB1
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CACNB1
Gene Accession No. P54283, Q02641, Q8R3Z5
Gene Alias CAB1, CACNB1, CACNLB1, CCHLB1
Gene Symbols Cacnb1
Host Species Rabbit
Immunogen CACNB1 Fusion Protein Ag3693
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12295, 50688, 782
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.