Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNB2 Antibody (N8b/1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147607H
Description
CACNB2 Monoclonal specifically detects CACNB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CACNB2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CAB2, CACNLB2, Calcium channel voltage-dependent subunit beta 2, calcium channel, voltage-dependent, beta 2 subunit, CAVB2, FLJ23743, Lambert-Eaton myasthenic syndrome antigen B, myasthenic (Lambert-Eaton) syndrome antigen B, MYSB, voltage-dependent L-type calcium channel subunit beta-2 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 78 kDa. No cross reactivity against CavB1, CavB3, CavB4. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S8b-1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q8VGC3 | |
CACNB2 | |
Synthetic peptide amino acids 189-205 of rat CavBeta2 | |
0.1 mL | |
783 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction