Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB2 Polyclonal Antibody

CACNB2 Polyclonal Antibody

Supplier:  Thermo Scientific PA550636

Catalog No. PA550636


Only null left
Add to Cart

Description

Description

CACNB2 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CACNB2
Polyclonal
Unconjugated
Cacnb2
AW060387; brain calcium channel beta-subunit CaB2c (splice variant of the gene CaB2); CAB1; CAB2; Cacnb2; CACNLB1; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; calcium voltage-gated channel auxiliary subunit beta 2; cardiac calcium channel beta subunit; cardiac calcium channel beta-subunit CaB2a (splice variant of the gene CaB2; cardiac calcium channel beta-subunit CaB2b (splice variant of the gene CaB2); CAVB2; Cavbeta2; Cchb2; CCHLB1; lambert-Eaton myasthenic syndrome antigen B; L-type calcium channel beta subunit; L-type voltage-dependent calcium channel beta-2 subunit; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; voltage-dependent calcium channel beta2A subunit; voltage-dependent calcium channel beta2B subunit; voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Rabbit
Antigen Affinity Chromatography
RUO
116600, 12296, 783
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q08289, Q8CC27, Q8VGC3
Cacnb2
Synthetic peptide corresponding to a region derived from internal residues of human calcium channel, voltage-dependent, beta 2 subunit.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.