Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206369
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6369 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6369 Supplier Fabgennix Inc Supplier No. CCB2101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CACNB2 Polyclonal antibody specifically detects CACNB2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen CACNB2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Cacnb2
Gene Accession No. Q08289, Q8CC27, Q8VGC3
Gene Alias AW060387; brain calcium channel beta-subunit CaB2c (splice variant of the gene CaB2); CAB1; CAB2; Cacnb2; CACNLB1; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; calcium voltage-gated channel auxiliary subunit beta 2; cardiac calcium channel beta subunit; cardiac calcium channel beta-subunit CaB2a (splice variant of the gene CaB2; cardiac calcium channel beta-subunit CaB2b (splice variant of the gene CaB2); CAVB2; Cavbeta2; Cchb2; CCHLB1; lambert-Eaton myasthenic syndrome antigen B; L-type calcium channel beta subunit; L-type voltage-dependent calcium channel beta-2 subunit; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; voltage-dependent calcium channel beta2A subunit; voltage-dependent calcium channel beta2B subunit; voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 500-600 on rat voltage-dependent L-type calcium channel subunit beta-2 protein
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116600, 12296, 783
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.