Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNB3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299461F
Description
CACNB3 Polyclonal antibody specifically detects CACNB3 in Human samples. It is validated for Western BlotSpecifications
CACNB3 | |
Polyclonal | |
Western Blot | |
CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNB3. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
784 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction