Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNB3 Monoclonal Antibody (7D1)
GREENER_CHOICE

Catalog No. PIMA527734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527734 Supplier Invitrogen™ Supplier No. MA527734
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: human CACNB3-transfected 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7D1
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Cacnb3
Gene Accession No. P54284
Gene Alias Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3
Gene Symbols Cacnb3
Host Species Mouse
Immunogen Human CACNB3.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.