Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109280 Supplier Invitrogen™ Supplier No. PA5109280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Rat brain tissue lysate, SKOV-3, human uterus tissue, mouse ovary tissue.

CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Cacnb3
Gene Accession No. P54284
Gene Alias Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3
Gene Symbols Cacnb3
Host Species Rabbit
Immunogen Synthetic peptide within Human CACNB3 aa 435-484.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.