Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNG1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$499.50

Specifications

Antigen CACNG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168998
SDP
View Documents
Novus Biologicals
NBP168998
100 μL
Each of 1 for $499.50
Only null left
Add to Cart
 
Description

Description

CACNG1 Polyclonal specifically detects CACNG1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CACNG1
Polyclonal
Rabbit
O70578
786
Synthetic peptides corresponding to Cacng1 (calcium channel, voltage-dependent, gamma subunit 1) The peptide sequence was selected from the N terminal of Cacng1. Peptide sequence AVHNKDKSCEHVTPSGEKNCSYFRHFNPGESSEIFEFTTQKEYSISAAAI.
Primary
Western Blot
Unconjugated
RUO
CACNLGDihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, calcium channel, voltage-dependent, gamma subunit 1, L-type calcium channel gamma polypeptide, neuronal dihydropyridine-sensitive calcium channel gamma subunit, voltage-dependent calcium channel gamma-1 subunit
CACNG1
IgG
25 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.