Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNG1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CACNG1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2137
![]() ![]() |
Proteintech Group Inc
183891AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Voltage-dependent calcium channels are essential for the release of neurotransmitters. L-type (long lasting current) voltage-dependent calcium channels are composed of four subunits: an alpha1 subunit, a beta subunit, a gamma subunit and an alpha2 delta subunit. The beta subunit is encoded by four genes, designated beta1-beta4, all of which contribute to the diversity of calcium currents and are involved in membrane trafficking of the alpha1 subunit. L-type Ca ++ CP gamma1, also known as CACNLG or CACNG1, is a 222 amino acid multi-pass membrane protein belonging to the PMP-22/EMP/MP20 family. Expressed in skeletal muscle, L-type Ca ++ CP gamma1 is a subunit of the dihydropyridine (DHP) sensitive calcium channel and may play a role in excitation-contraction coupling. L-type Ca++ CP gamma1 is considered a novel marker for maligt hyperthermia susceptibility (MHS), an autosomal domit disorder of skeletal muscle which manifests as a life-threatening hypermetabolic crisis triggered by commonly used inhalation anaesthetics and depolarizing muscle relaxants.Specifications
CACNG1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q06432 | |
786 | |
CACNG1 Fusion Protein Ag12918 | |
Primary | |
-20°C | |
CACNG1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CACNG1, CACNLG | |
CACNG1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title