Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNG2/4/8 Monoclonal Antibody (N245/36), APC
GREENER_CHOICE

Catalog No. PIMA545584
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545584 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545584 Supplier Invitrogen™ Supplier No. MA545584
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45584 was sufficient for detection of TARP Gamma2/4/8 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 35-55kDa. Does not cross-react with TARPGamma3. This antibody was formerly sold as clone S245-36.

Voltage-dependent calcium channel gamma-2 subunit, TARPGamma2 or Stargazin is encoded by the gene CACNG2 and is the transmembrane gamma subunit of the L-type calcium channel. Stargazin is in involved in regulation and trafficking of the ligand gated ion channel AMPA receptors. It is thought to play a role in learning and memory. Diseases associated with this gene include forms of deafness, Mental Retardation and childhood absnce epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG2/4/8
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N245/36
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Cacng8
Gene Accession No. O88602, Q71RJ2, Q8VHW2, Q8VHW5, Q8VHW9, Q8WXS5, Q9JJV4, Q9UBN1, Q9Y698
Gene Symbols Cacng2, Cacng4, Cacng8
Host Species Mouse
Immunogen Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10369, 12300, 140725, 140729, 27092, 54377, 59283, 81905, 84347
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.