Learn More
Invitrogen™ CACNG2/4/8 Monoclonal Antibody (N245/36), PerCP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545586
Description
1 μg/mL of MA5-45586 was sufficient for detection of TARP Gamma2/4/8 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 35-55kDa. Does not cross-react with TARPGamma3. This antibody was formerly sold as clone S245-36.
Voltage-dependent calcium channel gamma-2 subunit, TARPGamma2 or Stargazin is encoded by the gene CACNG2 and is the transmembrane gamma subunit of the L-type calcium channel. Stargazin is in involved in regulation and trafficking of the ligand gated ion channel AMPA receptors. It is thought to play a role in learning and memory. Diseases associated with this gene include forms of deafness, Mental Retardation and childhood absnce epilepsy.
Specifications
CACNG2/4/8 | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
O88602, Q71RJ2, Q8VHW2, Q8VHW5, Q8VHW9, Q8WXS5, Q9JJV4, Q9UBN1, Q9Y698 | |
Mouse | |
Protein G | |
RUO | |
10369, 12300, 140725, 140729, 27092, 54377, 59283, 81905, 84347 | |
4°C | |
Liquid |
Western Blot, Immunocytochemistry | |
N245/36 | |
PerCP | |
Cacng8 | |
Cacng2, Cacng4, Cacng8 | |
Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.