Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacng2/Cacng4/Cacng8 Mouse anti-Rat, HRP, Clone:S245-36, Abnova™

Catalog No. 89334109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-109 Supplier Abnova Corporation Supplier No. MAB17337
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant rat Cacng2/Cacng4/Cacng8.

Specifications

Antigen Cacng2/Cacng4/Cacng8
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S245-36
Conjugate HRP
Description calcium channel, voltage-dependent, gamma subunit 2
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Cacng2
Gene Alias Ipr328
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 203-323 at C-terminus of rat Cacng2.
Quantity 100 μg
Gene ID (Entrez) 84347, 140725, 140729
Target Species Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.