Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568376 Supplier Invitrogen™ Supplier No. PA568376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

Calcium Channel gamma-3 is a member of voltage dependent calcium channels (VDCCs) which allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal, and important in the genesis of synaptic transmission in discrete hippocampal fields. Calcium Channel gamma-3 is a type I transmembrane AMPA receptor regulatory protein (TARP). TARPs regulate both trafficking and channel gating of the AMPA receptors. Further, Calcium Channel gamma-3 is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family. Calcium Channel gamma-3 gene dysfunction is a susceptibility locus for childhood absence epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG3
Applications Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Cacng3
Gene Accession No. O60359, Q9JJV5
Gene Alias Cacng3; CACNLB3; calcium channel, voltage-dependent, gamma subunit 3; calcium voltage-gated channel auxiliary subunit gamma 3; CaVgamma3; neuronal voltage-gated calcium channel gamma-3 subunit; TARP gamma-3; transmembrane AMPAR regulatory protein gamma-3; Voltage-dependent calcium channel gamma-3 subunit; voltage-gated calcium channel gamma subunit
Gene Symbols Cacng3
Host Species Rabbit
Immunogen Full length Fusion protein of human Calcium Channel gamma-3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10368, 54376
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.