Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNG4 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299568MFV450
Description
CACNG4 Polyclonal antibody specifically detects CACNG4 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
CACNG4 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
calcium channel, voltage-dependent, gamma subunit 4, MGC11138, MGC24983, neuronal voltage-gated calcium channel gamma-4 subunit, TARP gamma-4, transmembrane AMPAR regulatory protein gamma-4, voltage-dependent calcium channel gamma-4 subunit | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNG4. | |
0.1 mL | |
Neuroscience, Signal Transduction | |
27092 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction