Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CACNG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566769
Description
Immunogen sequence: TLSREPLKVTT AASYSPDQEA SFLQVHDFFQ QDLKEGFHVS MLNRRTTPV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
The expression of multiple classes of voltage dependent calcium channels (VDCC's) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC's exist before birth which is consistent with a role in migration, most N-Type VDCC's subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications
CACNG4 | |
Polyclonal | |
Unconjugated | |
Cacng4 | |
AI413107; AW491861; CACNG4; calcium channel, voltage-dependent, gamma subunit 4; calcium voltage-gated channel auxiliary subunit gamma 4; CaV gamma4; MGC11138; MGC24983; neuronal voltage-gated calcium channel gamma-4 subunit; TARP gamma-4; transmembrane AMPAR regulatory protein gamma-4; Voltage-dependent calcium channel gamma-4 subunit | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27092 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBN1 | |
Cacng4 | |
Recombinant Human Calcium Channel gamma-4. Recombinant protein control fragment (Product #RP-107426). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction