Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109272 Supplier Invitrogen™ Supplier No. PA5109272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: MCF-7, SH-SY-5Y, rat testis tissue, rat brain tissue.

The expression of multiple classes of voltage dependent calcium channels (VDCC's) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC's exist before birth which is consistent with a role in migration, most N-Type VDCC's subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Cacng4
Gene Accession No. Q8VHW9, Q9UBN1
Gene Alias AI413107; AW491861; CACNG4; calcium channel, voltage-dependent, gamma subunit 4; calcium voltage-gated channel auxiliary subunit gamma 4; CaV gamma4; MGC11138; MGC24983; neuronal voltage-gated calcium channel gamma-4 subunit; TARP gamma-4; transmembrane AMPAR regulatory protein gamma-4; Voltage-dependent calcium channel gamma-4 subunit
Gene Symbols Cacng4
Host Species Rabbit
Immunogen Synthetic peptide within Human Cacng4 aa 189-238.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140725, 27092
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.