Learn More
Invitrogen™ CACNG5 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | CACNG5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Cacng5 |
| Gene Accession No. | Q9UF02 |
| Gene Alias | Cacng5; calcium channel, voltage-dependent, gamma subunit 5; calcium voltage-gated channel auxiliary subunit gamma 5; neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; transmembrane AMPAR regulatory protein gamma-5; voltage dependent calcium channel gamma 5 subunit; voltage-dependent calcium channel gamma-5 subunit |
| Gene Symbols | Cacng5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.