Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CACNG6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566028
Description
Immunogen sequence: NTYKANGSAVC EAAHLGLWKA CTKRLWQADV PVDRDTCGPA ELPGEANCTY FKFFTTGENA RIFQRTTK Highest antigen sequence identity to the following orthologs - mouse 91%, rat 90%.
Voltage-dependent calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of two known gamma subunit proteins. This particular gamma subunit is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two family members that function as transmembrane AMPA receptor regulatory proteins (TARPs). Alternative splicing results in multiple transcript variants. Variants in this gene have been associated with aspirin-intolerant asthma.
Specifications
CACNG6 | |
Polyclonal | |
Unconjugated | |
CACNG6 | |
2310033H20Rik; AW050150; CACNG6; calcium channel, voltage-dependent, gamma subunit 6; calcium voltage-gated channel auxiliary subunit gamma 6; neuronal voltage-gated calcium channel gamma-6 subunit; Voltage-dependent calcium channel gamma-6 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXT2 | |
CACNG6 | |
Recombinant Human Calcium Channel 6. Recombinant protein control fragment (Product #RP-106718). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction