Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548249
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548249 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548249 Supplier Invitrogen™ Supplier No. PA548249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with rat based on sequence homology.

Regulates the trafficking and gating properties of AMPA- selective glutamate receptors (AMPARs). Promotes their targeting to the cell membrane and synapses and modulates their gating properties by slowing their rates of activation, deactivation and desensitization and by mediating their resensitization. Does not show subunit-specific AMPA receptor regulation and regulates all AMPAR subunits. Thought to stabilize the calcium channel in an inactivated (closed) state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CACNG8
Gene Accession No. Q8VHW2, Q8WXS5
Gene Alias CACNG6; Cacng8; calcium channel, voltage-dependent, gamma subunit 8; calcium voltage-gated channel auxiliary subunit gamma 8; CaVGamma8; mir-935; Neuronal voltage-gated calcium channel gamma-8 subunit; TARP gamma 8; TARP gamma-8; Transmembrane AMPAR regulatory protein gamma-8; voltage-dependent calcium channel gamma-8 subunit
Gene Symbols CACNG8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 90-119 amino acids from the N-terminal region of human CACNG8.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59283, 81905
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.