Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cactin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Cactin |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Cactin Polyclonal specifically detects Cactin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cactin | |
Polyclonal | |
Rabbit | |
Human | |
Q8WUQ7 | |
58509 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TGFEWNKYNQTHYDFDNPPPKIVQGYKFNIFYPDLIDKRSTPEYFLEACADNKDFAILRFHAGPPYEDIAFKIVNREWEYSHRHGFRCQFANGIFQLWFH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C19orf29, cactin, cactin homolog, cactin, spliceosome C complex subunit, chromosome 19 open reading frame 29, FLJ17482, FLJ59622, fSAPc, functional spliceosome-associated protein c, NY REN 24 antigen, NY-REN-24, NY-REN-24 antigen | |
CACTIN | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title