Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACTIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523500
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523500 Supplier Invitrogen™ Supplier No. PA523500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Bacteria, Mosquito, Mouse, Pufferfish, Rat, Wild Turkey, Zebrafish.

Cactin is a novel negative regulator of TLR signaling and reveals its capacity to target MHC Class III genes at the molecular and functional level. It is localized to the nucleus, which is critical for manifesting its inhibitory effects on TLR signaling. Experimental evidences show overexpression of Cactin suppresses TLR-induced activation of NF-kB and interferon-regulatory factor transcription factors and induction of TLR-responsive genes. It is also reported Cactin interacts with IkB-like protein and targets other proteins that are encoded by genes in the MHC Class III region of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACTIN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene CACTIN
Gene Accession No. Q8WUQ7
Gene Alias 2510012J08Rik; C19orf29; C20H19orf29; CACTIN; cactin, spliceosome C complex subunit; fSAPc; functional spliceosome-associated protein c; NY REN 24 antigen; NY-REN-24; NY-REN-24 antigen; renal carcinoma antigen NY-REN-24; RGD1563634
Gene Symbols CACTIN
Host Species Rabbit
Immunogen A portion of amino acid 460-510 of human Cactin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100521180, 485055, 58509
Target Species Human, Canine, Monkey, Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.