Learn More
Invitrogen™ CACTIN Polyclonal Antibody

Description
Immunogen sequence: TGFEWNKYNQ THYDFDNPPP KIVQGYKFNI FYPDLIDKRS TPEYFLEACA DNKDFAILRF HAGPPYEDIA FKIVNREWEY SHRHGFRCQF ANGIFQLWFH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
Specifications
| Antigen | CACTIN |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CACTIN |
| Gene Accession No. | Q8WUQ7 |
| Gene Alias | 2510012J08Rik; C19orf29; C20H19orf29; CACTIN; cactin, spliceosome C complex subunit; fSAPc; functional spliceosome-associated protein c; NY REN 24 antigen; NY-REN-24; NY-REN-24 antigen; renal carcinoma antigen NY-REN-24; RGD1563634 |
| Gene Symbols | CACTIN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.