Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAD Antibody (SR1408), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CAD |
---|---|
Clone | SR1408 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:80, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
CAD Monoclonal antibody specifically detects CAD in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
CAD | |
Western Blot 1:500-1:1000, Flow Cytometry 1:80, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAD protein, CAD trifunctional protein, carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, anddihydroorotase, EC 2.1.3.2, EC 3.5.2.3, EC 6.3.5, EC 6.3.5.5, multifunctional protein CAD | |
A synthesized peptide derived from human CAD (Uniprot #: P27708) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1408 | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
790 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title