Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597015 Supplier Invitrogen™ Supplier No. PA597015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LERLGPGKGE VRPELGSRQD VEALWENMAV IDCFASDHAP HTLEEKCGSR PPPGFPGLET MLPLLLTAVS EGRLSLDDLL QRLHHNPRRI FHLPPQEDTY VEVDLEHEWT IPSHMPFSKA HWTPFEGQKV KGTVRRVVLR GEVAYIDGQV LVPPGYGQDV RKWPQGAVPQ LPPSAPATSE MTTTPERPRR GIPGLPDGRF HLPPRIHRAS DPGLPAEEPK EKSSRKVAEP ELMGTPDGTC YPPPPVPRQA S; Positive Samples: SW620, HepG2, HeLa, HL-60; Cellular Location: Cytoplasm, Nucleus.

The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAD
Gene Accession No. B2RQC6, P27708
Gene Alias 2410008J01Rik; 38E.19; anon-WO2004063362.83; Aspartate carbamoyltransferase; AU018859; cad; CAD protein; CAD trifunctional protein; cad-PA; cad-PB; cad-PC; carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase; carbamoylphosphate synthetase 2/aspartate transcarbamylase/dihydroorotase; carbamyl phosphatate synthetase 2; caudal; cb456; cd; CDG1Z; CG1759; CG1759-PA; CG1759-PB; CG1759-PC; Cpad; Dihydroorotase; Dmel\CG1759; Dmel_CG1759; Glutamine-dependent carbamoyl-phosphate synthase; Homeotic protein caudal; LD29596p; multifunctional protein CAD; S67; si:dkey-221h15.3; wu:fc30c12; wu:fc33d01; wu:fc67g02
Gene Symbols CAD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1650-1900 of human CAD (NP_0043322).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24240, 69719, 790
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.