Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
Shop All Cadence Science Inc. Products

Click to view available options
Diameter (English) Ball:
0.074 in.
0.078 in.
0.094 in.
0.11 in.
0.114 in.
Gauge:
14G
15G
17G
18G
20G
Length (English):
1 in.
1.2 in.
1.5 in.
2 in.
3 in.
3.1 in.
3.3 in.
4.3 in.
Description
- Nontoxic, nonpyrogenic
- PTFE tubing is flexible and softer than other types of tubing
- Low friction protects delicate organs
- Translucent for easy confirmation of reagent delivery
- Plastic luer hubs; soft silicone tips
- Sterile, packaged in pouches
- Five needles per pouch, 20 pouches per box
Specifications
Specifications
| Product Type | Soft, Flexible Animal Feeding Needle |
| Material | PTFE |
| Disposable | Single-use |
| For Use With (Application) | For critical applications requiring sterile, flexible, single use feeding needles |
| Gauge | 18G |
| Sterility | Sterile |
| Length (English) | 1.2 in. |
| Length (Metric) | 0.3 cm |
| Diameter (English) Ball | 0.078 in. |
| Diameter (Metric) Ball | 0.02 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction