Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-11 Antibody (1A5), R&D Systems™
SDP

Catalog No. NB780650 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB780650 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB780650 Supplier R&D Systems Supplier No. H00001009M04

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Cadherin-11 Monoclonal antibody specifically detects Cadherin-11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Functional Assay

Specifications

Antigen Cadherin-11
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone 1A5
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Functional
Formulation In 1x PBS, pH 7.4
Gene Accession No. P55287
Gene Alias CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin
Host Species Mouse
Immunogen CDH11 (P55287, 54 a.a. ~ 612 a.a) partial recombinant protein with Fc tag at C terminal. MW of the Fc tag alone is 50 KDa. 0
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1009
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.