Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-11 Antibody (667039), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB17901M
Description
Cadherin-11 Monoclonal antibody specifically detects Cadherin-11 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Cadherin-11 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
667039 | |
Flow Cytometry, CyTOF-ready | |
CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin | |
Mouse myeloma cell line NS0-derived recombinant human Cadherin-11, Phe23-Thr617, Accession # AAA35622 | |
0.1 mL | |
Cancer | |
1009 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction