Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-11 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP152045

Catalog No. NBP152045


Only null left
Add to Cart

Description

Description

Cadherin-11 Polyclonal specifically detects Cadherin-11 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Cadherin-11
Polyclonal
Western Blot 0.5 - 1 ug/ml, Peptide ELISA Detection limit 1:2000
NP_001788.2
CDH11
Peptide with sequence C-ERRGHLRPSFHG corresponding to internal region according to NP_001788.2.
0.1 mg
Cancer
1009
Human
IgG
Western Blot, ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin
Goat
Affinity Purified
RUO
Primary
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only