Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-12 Antibody (343621), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2240M
Description
Cadherin-12 Monoclonal antibody specifically detects Cadherin-12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Cadherin-12 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1010 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
343621 | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, FLJ34857, N-cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12, Met1-Pro609, Accession # P55289 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction