Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Antibody (7C9A10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306525PE
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human samples. It is validated for ELISA, ELISASpecifications
Cadherin-16 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
1014 | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Sandwich ELISA | |
7C9A10 | |
ELISA, Sandwich ELISA Detection | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-16 (Accession#: NP_004053.1; Met 1-Ala 786). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction