Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Antibody (CDH16/1071), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247745MFV450
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Flow CytometrySpecifications
Cadherin-16 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
1014 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
CDH16/1071 | |
Flow Cytometry | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Recombinant full-length human Cadherin-16 protein (Uniprot: O75309) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction