Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Antibody (CDH16/7027R), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314213JF646
This item is not returnable.
View return policy
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cadherin-16 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1014 | |
Store at 4°C in the dark. | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CDH16/7027R | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Recombinant human full-length Cadherin-16 protein (Uniprot: O75309 ) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction