Learn More
Cadherin-16 Antibody, FITC, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP299664F
Catalog No. NB062359
Only null left
Add to Cart
Description
Description
Cadherin-16 Polyclonal specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Cadherin-16
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Produced in rabbits immunized with purified, recombinant Cadherin-16 (NP_004053.1; Met1-Ala786).
0.1 mL
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS
Rabbit
Affinity purified
RUO
1014
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.