Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Antibody (rCDH16/1071), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308881AF700
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
Cadherin-16 | |
Monoclonal | |
Alexa Fluor 700 | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Recombinant full-length human Cadherin-16 protein (Uniprot: O75309) | |
100 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
rCDH16/1071 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
1014 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction