Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-17 Antibody (029), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289891B
Description
Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cadherin-17 | |
Monoclonal | |
Biotin | |
cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cadherin-17 (Q12864-1; Met1-Met787). | |
0.1 mL | |
Cancer | |
1015 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
29 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human Cad8 Human E-Cad Human Cad5 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction